Merck
  • Determination of nickel species in stack emissions from eight residual oil-fired utility steam-generating units.
Determination of nickel species in stack emissions from eight residual oil-fired utility steam-generating units.
Environmental science & technology (2011-06-30)
Frank E Huggins, Kevin C Galbreath, Kurt E Eylands, Lisa L Van Loon, Jeremy A Olson, Edward J Zillioux, Stephen G Ward, Paul A Lynch, Paul Chu
ZUSAMMENFASSUNG
XAFS spectroscopy has been used to determine the Ni species in particulate matter collected on quartz thimble filters in the stacks of eight residual (No. 6 fuel) oil-burning electric utility steam-generating units. Proper speciation of nickel in emitted particulate matter is necessary to correctly anticipate potential health risks. Analysis of the spectroscopic data using least-squares linear combination methods and a newly developed method specific for small quantities of Ni sulfide compounds in such emissions show that potentially carcinogenic Ni sulfide compounds are absent within the detection limits of the method (≤ 3% of the total Ni) in the particulate matter samples investigated. In addition to the major nickel sulfate phase (NiSO(4)·6H(2)O), lesser amounts of (Ni,Mg)O and/or NiFe(2)O(4) were also identified in most emission samples. On the basis of the results from these emission characterization studies, the appropriateness of the U.S. Environmental Protection Agency's assumption that the Ni compound mixture emitted from residual oil-fired power plants is 50% as carcinogenic as nickel subsulfide (Ni(3)S(2)) should be re-evaluated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nickelsulfid, 99.7% trace metals basis, −200 mesh