Merck
  • Synthesis and biological activity of 2-hydroxynicotinoyl-serine-butyl esters related to antibiotic UK-3A.
Synthesis and biological activity of 2-hydroxynicotinoyl-serine-butyl esters related to antibiotic UK-3A.
Bioorganic & medicinal chemistry letters (2010-07-14)
Ade Arsianti, Muhammad Hanafi, Endang Saepudin, Tsumoru Morimoto, Kiyomi Kakiuchi
ZUSAMMENFASSUNG
Novel 2-hydroxynicotinoyl-serine-butyl esters have been synthesized. Three-step reactions from L-serine by esterification with n-butanol, amidation with 2-hydroxynicotinic acid and esterification with the corresponding carboxylic acids gave AD-1, AD-2 and AD-3. The toxicity level of esters were determined by Brine shrimp assay, and antibiotic activity was tested against Escherichia coli, Bacillus subtilis, Staphylococcus aureus and Candida albicans. AD-3 showed greater activity as a growth inhibitor of B. subtilis and S. aureus compared to Antimycin A(3).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Hydroxypyridin-3-carbonsäure, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.