Merck
  • 2',3'-isopropylidene group, a molecular scaffold to study the activity of adenosine and adenylate deaminase on adenosine analogues modified in the ribose moiety.
2',3'-isopropylidene group, a molecular scaffold to study the activity of adenosine and adenylate deaminase on adenosine analogues modified in the ribose moiety.
Nucleosides, nucleotides & nucleic acids (2007-12-11)
Pierangela Ciuffreda, Laura Alessandrini, Enzo Santaniello
ZUSAMMENFASSUNG
2 ',3 '-Isopropylidene group can be used as a molecular scaffold for the introduction of modifications at 5 ' and 1 ' positions of adenosine and these modified nucleosides are used to evaluate the biocatalytic activity of adenosine and adenylate deaminase.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2′,3′-O-Isopropylidenadenosin, 98%