Merck
  • Tcf-1-mediated transcription in T lymphocytes: differential role for glycogen synthase kinase-3 in fibroblasts and T cells.
Tcf-1-mediated transcription in T lymphocytes: differential role for glycogen synthase kinase-3 in fibroblasts and T cells.
International immunology (1999-04-30)
F J Staal, B M Burgering, M van de Wetering, H C Clevers
ZUSAMMENFASSUNG
Beta-catenin is the vertebrate homolog of the Drosophila segment polarity gene Armadillo and plays roles in both cell-cell adhesion and transduction of the Wnt signaling cascade. Recently, members of the Lef/Tcf transcription factor family have been identified as protein partners of beta-catenin, explaining how beta-catenin alters gene expression. Here we report that in T cells, Tcf-1 also becomes transcriptionally active through interaction with beta-catenin, suggesting that the Wnt signal transduction pathway is operational in T lymphocytes as well. However, although Wnt signals are known to inhibit the activity of the negative regulatory protein kinase glycogen synthase kinase-3beta (GSK-3beta), resulting in increased levels of beta-catenin, we find no evidence for involvement of GSK-3beta in Tcf-mediated transcription in T cells. That is, a dominant negative GSK-3beta does not specifically activate Tcf transcription and stimuli (lithium or phytohemagglutinin) that inhibit GSK-3beta activity also do not activate Tcf reporter genes. Thus, inhibition of GSK-3beta is insufficient to activate Tcf-dependent transcription in T lymphocytes. In contrast, in C57MG fibroblast cells, lithium inactivates GSK-3beta and induces Tcf-controlled transcription. This is the first demonstration that lithium can alter gene expression of Tcf-responsive genes, and points to a difference in regulation of Wnt signaling between fibroblasts and lymphocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
TCF-Reporter-Plasmid-Kit, Set of transfection grade T cell factor (TCF) reporter plasmids for use in TOPFlash and FOPFlash wnt/b-catenin activity assays.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.