Merck
Artificial leaf device for solar fuel production.
Faraday discussions (2012-04-05)
Yutaka Amao, Naho Shuto, Kana Furuno, Asami Obata, Yoshiko Fuchino, Keiko Uemura, Tsutomu Kajino, Takeshi Sekito, Satoshi Iwai, Yasushi Miyamoto, Masatoshi Matsuda
ZUSAMMENFASSUNG
Solar fuels, such as hydrogen gas produced from water and methanol produced from carbon dioxide reduction by artificial photosynthesis, have received considerable attention. In natural leaves the photosynthetic proteins are well-organized in the thylakoid membrane. To develop an artificial leaf device for solar low-carbon fuel production from CO2, a chlorophyll derivative chlorin-e6 (Chl-e6; photosensitizer), 1-carboxylundecanoyl-1'-methyl-4,4'-bipyrizinium bromide, iodide (CH3V(CH2)9COOH; the electron carrier) and formate dehydrogenase (FDH) (the catalyst) immobilised onto a silica-gel-based thin layer chromatography plate (the Chl-V-FDH device) was investigated. From luminescence spectroscopy measurements, the photoexcited triplet state of Chl-e6 was quenched by the CH3V(CH2)9COOH moiety on the device, indicating the photoinduced electron transfer from the photoexcited triplet state of Chl-e6 to the CH3V(CH2)9COOH moiety. When the CO2-saturated sample solution containing NADPH (the electron donor) was flowed onto the Chl-V-FDH device under visible light irradiation, the formic acid concentration increased with increasing irradiation time.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formiatdehydrogenase aus Candida boidinii, lyophilized powder, 5.0-15.0 units/mg protein