Merck
  • Effects of the novel angiotensin II receptor type I antagonist, fimasartan on myocardial ischemia/reperfusion injury.
Effects of the novel angiotensin II receptor type I antagonist, fimasartan on myocardial ischemia/reperfusion injury.
International journal of cardiology (2013-05-07)
Jin Han, Sung-Ji Park, Vu Thi Thu, Sung-Ryul Lee, Le Thanh Long, Hyoung Kyu Kim, Nari Kim, Seung Woo Park, Eun-Seok Jeon, Eun-Ji Kim, Chang-Hwan Yoon, Goo-Young Cho, Dong-Ju Choi
ZUSAMMENFASSUNG
The aim of this study was to investigate the cardioprotective effect of fimasartan, a newly developed angiotensin II receptor type I blocker (ARB), against myocardial ischemia/reperfusion (I/R) injury and to identify the mechanism by which it reduces mitochondrial damage. Fimasartan was administered intravenously to Sprague-Dawley rats (3mg/kg), cardiomyocytes (50 μM), and H9c2 cells (50 μM) before ischemia or hypoxia. Myocardial infarction (MI), echocardiograms, DNA fragmentation, terminal deoxynucleotidyl transferase-mediated dUTP in situ nick-end labeling, immunoblotting, oxygen consumption, confocal microscopic appearance, and L-type Ca(2+) current (ICa,L) were then assessed. Fimasartan pretreatment remarkably reduced the rate of MI and improved cardiac performance well after I/R (n = 9/group). Fimasartan also reduced apoptotic cell death both in vivo and in hypoxia/reoxygenation (H/R)-treated H9c2 cells (n = 5~8/group). H/R-induced mitochondrial O2(-) production and collapse of membrane potential were markedly attenuated in fimasartan-treated cardiomyocytes (n = 4 ~ 6/group). Additionally, mitochondrial Ca(2+) overload during reoxygenation was suppressed by fimasartan (n = 4~6/group), and this was found to be possibly related to the inhibition of ICa,L and mitochondrial Ca(2+) uniporter. Furthermore, fimasartan pretreatment increased phosphorylations of Akt and glycogen synthase kinase-3β (n = 5 ~ 7/group), decreased pro-apoptotic p53 levels, and increased anti-apoptotic Bcl-2 levels (n = 4) during reperfusion. Fimasartan preconditioning has the potential to modulate Bcl-2 and suppress I/R-induced Ca(2+) overload by inhibiting ICa,L and MCU. These beneficial effects could prevent the mitochondrial dysfunction and apoptosis accompanied by I/R.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fimasartan, ≥98% (HPLC)