Merck
Synthesis of Plant Phenol-derived Polymeric Dyes for Direct or Mordant-based Hair Dyeing.
Journal of visualized experiments : JoVE (2016-12-09)
Kyung Min Im, Jong-Rok Jeon
ZUSAMMENFASSUNG
Effective hair dyeing through in situ incubation of keratin hair with the products of fungal laccase-catalyzed polymerization of plant phenols has been previously demonstrated. However, the dyeing process takes a long time to complete compared to commercial hair-dyeing products. To overcome this bottleneck, pre-synthesized polymeric products of the oxidative reaction of Trametes versicolor laccase on catechin and catechol, either with or without mordant agents (e.g., FeSO4), were here employed to achieve permanent keratin hair dyeing in various colors and shades. The laccase action in acidic sodium acetate buffer led to a deep black coloration after coupling reactions between the plant phenols. The colored dye products were then desalted and concentrated with ultrafiltration. The dyes, with or without mordant agents, caused a significant increase in ΔE values (i.e., color difference value) in gray human hair within 2.5 hours. In addition, different keratin colors and shades were induced depending upon the mordanting and pH changes. The dyed hair also exhibited a strong resistance to detergent treatments, indicating that our methods can give rise to permanent hair dyeing. Overall, our work has provided novel insight into developing eco-friendly hair-dyeing methods as alternatives to commercial toxic diamine-based dyes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
Ultrafiltrationsscheiben, 5 kDa NMW, Ultracel®, filter diam. 63.5 mm