Merck
9-Substituted berberine derivatives as G-quadruplex stabilizing ligands in telomeric DNA.
Bioorganic & medicinal chemistry (2007-06-19)
Wan-Jin Zhang, Tian-Miao Ou, Yu-Jing Lu, Ying-Yu Huang, Wei-Bin Wu, Zhi-Shu Huang, Jin-Lin Zhou, Kwok-Yin Wong, Lian-Quan Gu
ZUSAMMENFASSUNG
The interaction of berberine and its 9-substituted derivatives with human telomeric DNA d[G(3)(T(2)AG(3))(3)](telo21) has been investigated via CD spectroscopy, fluorescence spectroscopy, PCR-stop assay, competitive dialysis, and telomerase repeat amplification protocol (TRAP) assay. The results indicated that these semisynthesized compounds could induce and stabilize the formation of anti-parallel G-quadruplex of telomeric DNA in the presence or absence of metal cations. Compared with berberine, the 9-substituted derivatives exhibit stronger binding affinity with G-quadruplex and higher inhibitory activity for telomerase. Introduction of a side chain with proper length of methylene and terminal amino group to the 9-position of berberine would significantly strengthen the binding affinity with G-quadruplex, resulting in increasing inhibitory effects on the amplification of telo21 DNA and on the telomerase activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Berberin Chlorid-Form