Merck
Simultaneous analysis of thirteen phytohormones in fruits and vegetables by SPE-HPLC-DAD.
Food science and biotechnology (2020-10-23)
Song Zhu, Shang-Wei Chen, Yue Li
ZUSAMMENFASSUNG
Determination of phytohormones have attracted increasing attentions in food safety field. In this study, an efficient and quantitative method was developed which can simultaneously determinate thirteen phytohormones in fruits and vegetables using solid phase extraction (SPE) combined with high performance liquid chromatography-diode array detection (HPLC-DAD). The samples were extracted with 80% methanol containing 0.5% (V/V) formic acid, and the extracts were then concentrated and purified using primary secondary amine (PSA) and C18 tandem dual SPE cartridges. The analytes were separated on a Waters XBridge™ C18 column and eluated utilizing a gradient elution program of water and methanol. Mean recoveries of the thirteen analytes varied from 74.69 to 92.40%, with relative standard deviations < 3.57%. The limits of detection and quantitation were 0.005-0.018 mg/kg and 0.02-0.10 mg/kg, respectively. The phytohormones in kiwi fruit, strawberry, bean sprout, and green pepper were detected using the above method, respectively. Only the IAA content of 0.14 mg/kg was detected for the strawberry from a supermarket, which was lower than the prescribed limit in food safety standards (0.2 mg/kg).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Indol-3-essigsäure Natriumsalz, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
4-Chlorphenoxyessigsäure, ≥98.0% (T)