Merck
  • Color Stability of Fermented Mare's Milk and a Fermented Beverage from Cow's Milk Adapted to Mare's Milk Composition.
Color Stability of Fermented Mare's Milk and a Fermented Beverage from Cow's Milk Adapted to Mare's Milk Composition.
Foods (Basel, Switzerland) (2020-02-26)
Joanna Teichert, Dorota Cais-Sokolińska, Romualda Danków, Jan Pikul, Sylwia Chudy, Paulina Bierzuńska, Łukasz K Kaczyński
ZUSAMMENFASSUNG
Color is important for the consumer when making a purchase decision. Mare's milk and, thus, fermented mare's milk is little known to consumers. Thus, it is worth presenting research showing the extent of color change during the production and storage of mare's milk. Herein, we examined the range of color changes in mare's milk and cow's milks adapted to mare's milk composition. These samples were further fermented and stored for 3 weeks at 5 ± 1 °C. Starter cultures containing Streptococcus thermophilus and Lactobacillus delbrueckii subsp. bulgaricus were used for fermentation. Mare's milk reached the required pH of 4.5 during fermentation faster (255 min) than cow's milk (300 min). After fermentation, mare's milk compared to cow's milk and adapted cow's milk had lower titratable acidity (0.75%) and firmness (145. 6 |(g∙s)|). The water holding capacity (95.6%) and number of Lactobacillus (7.71 log CFU/mL) and Streptocococcus (7.20 log CFU/mL) in mare's and other's milks were the same. Mare's milk was furthest from the ideal white (WI) color, with its chrome (C*) being 1.5-times larger than cow's milk. However, fermented mare's milk was darker than the fermented adapted milk and cow's milk by 36% and 58%, respectively. Storage caused a decrease in the WI, C*, and yellowness index (YI). The fermented mare's milk color stability during production and storage was less than that of fermented cow's milk. After 3 weeks storage, it was observed that the titratable acidity increased to 1.05%, and the pH decreased to 4.3 in fermented mare's milk. The water holding capacity decreased but was still higher compared to fermented cow's milk.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
GranuCult® Prime MRS-Agar (nach DeMAN, ROGOSA und SHARPE), according to ISO 15214, for Lactobacillus spp., suitable for microbiology