Merck
  • Short-chain fatty acid mitigates adenine-induced chronic kidney disease via FFA2 and FFA3 pathways.
Short-chain fatty acid mitigates adenine-induced chronic kidney disease via FFA2 and FFA3 pathways.
Biochimica et biophysica acta. Molecular and cell biology of lipids (2020-02-18)
Daisuke Mikami, Mamiko Kobayashi, Junsuke Uwada, Takashi Yazawa, Kazuko Kamiyama, Kazuhisa Nishimori, Yudai Nishikawa, Sho Nishikawa, Seiji Yokoi, Hideki Kimura, Ikuo Kimura, Takanobu Taniguchi, Masayuki Iwano
ZUSAMMENFASSUNG
Short-chain fatty acids (SCFAs), including acetate, butyrate, and propionate, are produced when colonic bacteria in the human gastrointestinal tract ferment undigested fibers. Free fatty acid receptor 2 (FFA2) and FFA3 are G-protein-coupled receptors recently identified as SCFA receptors that may modulate inflammation. We previously showed through in vitro experiments that SCFAs activate FFA2 and FFA3, thereby mitigating inflammation in human renal cortical epithelial cells. This study used a murine model of adenine-induced renal failure to investigate whether or not SCFAs can prevent the progression of renal damage. We also examined whether or not these FFA2 and FFA3 proteins have some roles in this protective mechanism in vivo. Immunohistochemical analyses of mouse kidneys showed that FFA2 and FFA3 proteins were expressed mainly in the distal renal tubules and collecting tubules. First, we observed that the administration of propionate mitigated the renal dysfunction and pathological deterioration caused by adenine. Consistent with this, the expression of inflammatory cytokines and fibrosis-related genes was reduced. Furthermore, the mitigation of adenine-induced renal damage by the administration of propionate was significantly attenuated in FFA2-/- and FFA3-/- mice. Therefore, the administration of propionate significantly protects against adenine-induced renal failure, at least in part, via the FFA2 and FFA3 pathways. Our data suggest that FFA2 and FFA3 are potential new therapeutic targets for preventing or delaying the progression of chronic kidney disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumacetat-d3, 99 atom % D
Sigma-Aldrich
Butter-d7-säure, ≥98 atom % D, ≥98% (CP)