- Generation of an induced pluripotent stem cell line (TRNDi004-I) from a Niemann-Pick disease type B patient carrying a heterozygous mutation of p.L43_A44delLA in the SMPD1 gene.
Generation of an induced pluripotent stem cell line (TRNDi004-I) from a Niemann-Pick disease type B patient carrying a heterozygous mutation of p.L43_A44delLA in the SMPD1 gene.
Stem cell research (2019-04-23)
Amanda Baskfield, Rong Li, Jeanette Beers, Jizhong Zou, Chengyu Liu, Wei Zheng
PMID31009819
ZUSAMMENFASSUNG
Niemann-Pick disease type B (NPB) is a rare autosomal recessive lysosomal storage disease caused by mutations in the SMPD1 gene, which encodes for acid sphingomyelinase. A human induced pluripotent stem cell (iPSC) line was generated from dermal fibroblasts of a 1-year old male patient with NPB that has a heterozygous mutation of a p.L43_A44delLA of SMPD1 using non-integrating Sendai virus technique. This iPSC line offers a useful resource to study the disease pathophysiology and as a cell-based model for drug development to treat NPB.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Milli-Mark® Anti-Nanog-Alexa Fluor 488 Antibody, NT, Milli-Mark®, from rabbit