Merck
  • Effect of Vitis vinifera hydroalcoholic extract against oxaliplatin neurotoxicity: in vitro and in vivo evidence.
Effect of Vitis vinifera hydroalcoholic extract against oxaliplatin neurotoxicity: in vitro and in vivo evidence.
Scientific reports (2018-09-27)
Laura Micheli, Luisa Mattoli, Anna Maidecchi, Alessandra Pacini, Carla Ghelardini, Lorenzo Di Cesare Mannelli
ZUSAMMENFASSUNG
Oxaliplatin treatment is associated with the development of a dose-limiting painful neuropathy impairing patient's quality of life. Since oxidative unbalance is a relevant mechanism of oxaliplatin neurotoxicity, we assessed the potential antioxidant properties of Vitis vinifera extract in reducing oxaliplatin-induced neuropathy as a valuable therapeutic opportunity. A hydroalcoholic extract of Vitis vinifera red leaf was characterized and tested in primary rat astrocyte cells treated with oxaliplatin (100 μM). Oxaliplatin lethality in the human adenocarcinoma cell line HT-29 was evaluated in the absence and presence of the extract. In vivo, pain hypersensitivity was measured in a rat model of neuropathy induced by oxaliplatin and ex vivo molecular targets of redox balance were studied. Vitis vinifera extract (50 μg mL-1, 4 h incubation) significantly reduced the oxaliplatin-dependent superoxide anion increase and lipid peroxidation in rat astrocytes but did not interfere with the mortality elicited by oxaliplatin in HT-29 cancer cells. In oxaliplatin-treated rats, a repeated daily administration of the Vitis vinifera extract (300 mg kg-1, p.o.) significantly prevented mechanical and thermal hypersensitivity to noxious and non noxious stimuli. mRNA and protein levels of Nrf2 were normalized in spinal cord and DRGs. Moreover, in the spinal cord, the extract significantly decreased the activation of astrocytes. Vitis vinifera reduced oxidative damages and relieved pain without influencing oxaliplatin anti-cancer activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Superoxid-Dismutase Rind, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)