Fortfahren mit
Z757802
Sartorius filter pipette tips
volume range 5-300 μL, sterile, pack of 960 ea (10 racks of 96tips)
Synonym(e):
filter pipette tips, pipetting tips, pipettor tips
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Ihnen/SKUVerfügbarkeitPreis
960 ea
Warenkorb auf Verfügbarkeit prüfen
€ 193,00
Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41121601
Volume:
5—300 μL
Sterility:
sterile
Feature:
DNase free, RNase free, barrier, filter
Packaging:
pack of 960 ea (10 racks of 96tips)
Manufacturer/tradename:
Sartorius 790301F
€ 193,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
colorless , polypropylene (Virgin)
sterility
sterile
feature
DNase free, RNase free, barrier, filter
packaging
pack of 960 ea (10 racks of 96tips)
manufacturer/tradename
Sartorius 790301F
maximum volume
300 μL
volume range
5-300 μL
volume
5—300 μL
binding type
low binding surface
General description
The Sartorius filter tips of the Biohitfamily are made of polypropylene and feature a polyethylene filter barrier to prevent aerosol and liquid contamination. This filter helps protect against the rist of cross-contamination and reduces maintenance requirements of the pipette. The filter tips are suited for molecular biology, microbiology, cell culture and radioactive work.
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
  • Pipette foaming liquids like buffers and proteins
  • Pipette solvents
  • Reverse pipetting
  • Multiple dispensing functions of electronic pipettes
Other Notes
Browse all pipette tips
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z757799Z757772Z757764
description
volume range 5-300 μL, sterile, pack of 960 ea (10 racks of 96tips)
description
volume range 5-200 μL, sterile, pack of 960 ea (10 racks of 96 tips)
description
volume range 0.5-20 μL, sterile, pack of 960 ea (10 racks of 96 tips)
description
volume range 0.1-10 μL, sterile, pack of 960 ea (10 racks of 96 tips)
volume
5—300 μL
volume
5—200 μL
volume
0.5—20 μL
volume
0.1—10 μL
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
feature
DNase free, barrier, RNase free, filter
feature
DNase free, RNase free, barrier, filter
feature
DNase free, RNase free, barrier, filter
feature
DNase free, RNase free, barrier, filter
manufacturer/tradename
Sartorius 790301F
manufacturer/tradename
Sartorius 790201F
manufacturer/tradename
Sartorius 790021F
manufacturer/tradename
Sartorius 790011F
material
colorless , polypropylene (Virgin)
material
colorless , polypropylene (Virgin)
material
colorless , polypropylene (Virgin)
material
colorless , polypropylene (Virgin)
binding type
low binding surface
binding type
low binding surface
binding type
low binding surface
binding type
low binding surface
Still not finding the right product?
Explore all of our products under Sartorius filter pipette tips
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)