Direkt zum Inhalt
Merck

M183

Memantin -hydrochlorid

Synonym(e):

3,5-Dimethyl-1-adamantanamin -hydrochlorid, 3,5-Dimethyl-amantadin -hydrochlorid, 3,5-Dimethyltricyclo[3.3.1.13,7]decan-1-amin -hydrochlorid

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen


Über diesen Artikel

Empirische Formel (Hill-System):
C12H21N · HCl
CAS-Nummer:
Molekulargewicht:
215.76
EC Number:
255-219-6
UNSPSC Code:
41116107
MDL number:
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten

InChI key

LDDHMLJTFXJGPI-VXMZOEJHSA-N

InChI

1S/C12H21N.ClH/c1-10-3-9-4-11(2,6-10)8-12(13,5-9)7-10;/h9H,3-8,13H2,1-2H3;1H/t9-,10+,11-,12-;

SMILES string

Cl.C[C@]12CC3C[C@](C)(C1)CC(N)(C3)C2

format

neat

Suchen Sie nach ähnlichen Produkten? Aufrufen Leitfaden zum Produktvergleich

Lagerklasse

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen

Ramya Varatharajan et al.
Neurochemistry international, 90, 255-260 (2015-10-10)
Deep brain stimulation (DBS) of the nucleus accumbens (NAc) is an effective treatment option for those affected by obsessive compulsive disorder, who do not respond to pharmacological treatment strategies. Yet, little is known about the mechanism by which DBS achieve
Felice Iasevoli et al.
Progress in neuro-psychopharmacology & biological psychiatry, 54, 299-314 (2014-07-16)
A relevant role for dopamine-glutamate interaction has been reported in the pathophysiology and treatment of psychoses. Dopamine and glutamate may interact at multiple levels, including the glutamatergic postsynaptic density (PSD), an electron-dense thickening that has gained recent attention as a
Teresa A Barber et al.
Behavioural pharmacology, 26(4), 407-410 (2015-03-05)
Evidence of observational learning (social learning) is present in many species. One such task is the one-trial taste-avoidance task, in which Actor chicks peck a bead coated with an aversant substance. Observer chicks learn to avoid beads that are similar
Tetsuaki Hiyoshi et al.
Neuropharmacology, 85, 384-396 (2014-06-08)
Schizophrenic patients have been shown to exhibit abnormal cortical gamma band oscillation (GBO), which is thought to be related to the symptoms of schizophrenia, including cognitive impairment. Recently, non-competitive NMDA receptor (NMDAr) antagonists such as MK-801 and ketamine have been
Kazuoki Hirano et al.
Biochemical and biophysical research communications, 518(1), 161-170 (2019-08-23)
Chemical intervention of autophagy has been investigated in clinical trials for various age-related conditions such as sarcopenia and neurodegeneration. However, at present, no autophagy inducer has been established as a disease-modifying agent against neurodegenerative diseases. We screened a library consisting

Unser Team von Wissenschaftlern verfügt über Erfahrung in allen Forschungsbereichen einschließlich Life Science, Materialwissenschaften, chemischer Synthese, Chromatographie, Analytik und vielen mehr..

Setzen Sie sich mit dem technischen Dienst in Verbindung