Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 130,00 |
Über diesen Artikel
€ 130,00
material
acrylic , clear (workstation)
feature
holds 4 x pipettes and pipette tips
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS234631
L × W × H
244 mm × 165 mm × 246 mm
General description
Features and Benefits
- Acrylic workstation is opaque on sides with clear front and bin lids for easy content identification
- Holds up to four pipettes on top shelf
- Dual bins hold multiple size tips
- Self-closing lid provides dust-free storage
- Oversized lip allows for easy access when wearing gloves
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| material acrylic , clear (workstation) | material acrylic , clear | material acrylic , clear | material acrylic , blue |
| manufacturer/tradename Heathrow Scientific HS234631 | manufacturer/tradename Heathrow Scientific HS206204 | manufacturer/tradename Heathrow Scientific HS20620 | manufacturer/tradename Heathrow Scientific HS20613C |
| L × W × H 244 mm × 165 mm × 246 mm | L × W × H 199 mm × 172 mm × 206.4 mm | L × W × H 295.3 mm × 172 mm × 203.2 mm | L × W × H 295.3 mm × 172 mm × 203.2 mm |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| feature holds 4 x pipettes and pipette tips | feature to hold 4 pipette(s) | feature to hold 6 pipette(s) | feature to hold 6 pipette(s) |
Still not finding the right product?
Explore all of our products under Pipette Workstation
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?