Fortfahren mit
zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
100 ea | Warenkorb auf Verfügbarkeit prüfen | € 81,00 |
Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41121522
Manufacturer/tradename:
Heathrow Scientific 120690
Capacity:
50 mL
Material:
PVC , V-bottom bottom
Sterility:
non-sterile
€ 81,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
PVC , V-bottom bottom
sterility
non-sterile
packaging
pack of 100 (bulk packed)
manufacturer/tradename
Heathrow Scientific 120690
capacity
50 mL
General description
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Features and Benefits
- Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sealed in easy-to-open bags. No scissors needed to open
- Disposable
- Design Patent Pending
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| sterility non-sterile | sterility sterile; electron beam irradiated | sterility non-sterile | sterility non-sterile |
| capacity 50 mL | capacity 50 mL | capacity 50 mL | capacity 10 mL |
| manufacturer/tradename Heathrow Scientific 120690 | manufacturer/tradename Heathrow Scientific 120646 | manufacturer/tradename Heathrow Scientific 120645 | manufacturer/tradename Heathrow Scientific 120638 |
| material PVC , V-bottom bottom | material PVC , V-bottom bottom | material V-bottom bottom, polystyrene | material V-bottom bottom, polystyrene |
| packaging pack of 100 (bulk packed) | packaging pack of 100 (packed as 5 ea) | packaging pack of 100 (bulk packed) | packaging pack of 100 (bulk packed) |
Still not finding the right product?
Explore all of our products under Reagent Reservoir
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?