Merck
GN781289
Greiner microplate
384 well, conical bottom, black
Synonym(e):
384 well microplate, 384 well plate, cell culture plate
About This Item
UNSPSC-Code:
41121800
Materialien
black
Verpackung
pack of 100 ea
Hersteller/Markenname
Greiner 781289
Verfügbarkeit
available only in China
Größe
384 wells
Eignung
suitable for (cell culture)
Allgemeine Beschreibung
  • Greiner microplates are ideal for storing active agents, patient samples, DNA/RNA or stock cultures.
  • Polypropylene material can withstand a high temperature (-196 to +121°C) and are chemically resistant.
  • Nonpyrogenic plates are free of detectable DNase, RNase, human DNA.
  • Features alphanumeric well coding for identification.
  • Sealable with adhesive films and heat sealers for long-term and secure sealing.
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)