Merck
EP0030075161
Eppendorf® epT.I.P.S.® G Racks, Biopur®
size (50-1250 μL), L (103 mm), dark green, pack of 480 ea (5 trays of 96 tips)
About This Item
UNSPSC-Code:
41121600
Materialien
green (Tray)
Sterilität
sterile
Leistungsmerkmale
DNase free
Verpackung
pack of 480 ea (5 trays of 96 tips)
Hersteller/Markenname
Eppendorf® 0030075161
Länge
(103 mm)
Maximales Volumen
1.25 mL
Größe
(50-1250 μL)
Farbe
dark green
Rechtliche Hinweise
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG