Merck
C5002
Sigma-Aldrich
Z-Pro-Leu-Gly amide
About This Item
Empirische Formel (Hill-System):
C21H30N4O5
CAS-Nummer:
Molekulargewicht:
418.49
MDL-Nummer:
UNSPSC-Code:
12352200
PubChem Substanz-ID:
SMILES String
CC(C)CC(NC(=O)C1CCCN1C(=O)OCc2ccccc2)C(=O)NCC(N)=O