Fortfahren mit
BAF187450010
Bel-Art® Switch-Grid® Test Tube Rack
Holds 84 x 10-13 mm tubes, orange grid
Synonym(e):
tube holder, 187450010, F18745-0010, tube organiser, tube rack, tube storage
zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
NACRES:
NB.35
UNSPSC Code:
41122800
Manufacturer/tradename:
Bel-Art F18745-0010
Feature:
autoclavable, Holds 84 x 10-13 mm tubes
Material:
orange grid, polyoxymethylene
Packaging:
pack of 1 ea
material
orange grid, polyoxymethylene
feature
autoclavable, Holds 84 x 10-13 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18745-0010
L × W × H
31.8 cm × 11.1 cm × 6.7 cm
General description
1 RACK FRAME, 25 POSSIBLE TUBE SIZE COMBINATIONS
This innovative, split-frame rack holds up to two different size tubes in the same rack and allows users to quickly switch grids to hold other sizes as needed. Each rack frame includes a set of grids in one size. Grid sets in alternate sizes can be purchased as needed.
Features and Benefits
More compact and less expensive than purchasing multiple racks
Colored grid inserts correspond to a specific test tube size ranging from 10mm to 30mm in diameter
Three tier rack holds tubes securely in place
Rubber feet keep racks from sliding on benchtop, provide secure stacking while in storage
Chemical resistant polyoxymethylene plastic rack frame and grids can be cleaned with mild detergent, ethanol or isopropyl alcohol; autoclaved at 121°C (250°F).
Legal Information
Bel-Art is a registered trademark of Bel-Art Products, a division of SP
Switch-Grid is a registered trademark of Bel-Art Products, a division of SP
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
BAF187450045BAF188710000T6194
feature
autoclavable, Holds 84 x 10-13 mm tubes
feature
-
feature
to hold 120 x 13-16mm Tubes
feature
Holds 96 x 15 mL tubes
manufacturer/tradename
Bel-Art F18745-0010
manufacturer/tradename
Bel-Art F18745-0045
manufacturer/tradename
Bel-Art F18871-0000
manufacturer/tradename
Bel-Art F18765-0001
material
orange grid, polyoxymethylene
material
plastic
material
-
material
epoxy (coated steel)
L × W × H
31.8 cm × 11.1 cm × 6.7 cm
L × W × H
-
L × W × H
-
L × W × H
24.2 cm (9¹/₂ in.) × 17 cm (6¹¹/₁₆ in.) × 6.4 cm (2¹/ in.)
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
Bel-Art Switch-Grid Test Tube Rack 25 combinations
-
-
-
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)