Merck
AXYEV200NTRS
Corning® Axygen® Automation Tips for Tecan® Freedom Evo®, TeMo and Aquarius
volume (50 μL), round clear polypropylene tip, Non-Filtered, sterile
Synonym(e):
pipet tips
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.02
Materialien
round clear polypropylene tip
Sterilität
sterile
Verpackung
case of 3,840 tips
pack of 768 tips
pkg of 8 Wafers/Stack, 5 Stacks/Cs
Hersteller/Markenname
Corning EV-200-NTR-S
Volumen
(50 μL)
Rechtliche Hinweise
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Evo is a registered trademark of Corning, Inc.
Tecan is a registered trademark of Tecan Group AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)