Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
PTFE plunger tip
size
100 μL
feature
1710C, LT, N, RN, or TLL
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 1162-02
syringe volume
100 μL
compatibility
for use with Hamilton 1710 (C, LT, N, RN, or TLL)
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| compatibility for use with Hamilton 1710 (C, LT, N, RN, or TLL) | compatibility for use with Hamilton 1750 (LT, LTN, RN, or TLL) | compatibility for use with Hamilton 1705 (LT, RN, or TLL) | compatibility for use with Hamilton 1725 (LT, N, RN, or TLL) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| material PTFE plunger tip | material PTFE plunger tip | material PTFE plunger tip | material PTFE plunger tip |
| feature 1710C, LT, N, RN, or TLL | feature 1750 LT, LTN, RN, or TLL | feature 1705 LT, RN, or TLL | feature 1725LT, N, RN, or TLL |
| manufacturer/tradename Hamilton 1162-02 | manufacturer/tradename Hamilton 1169-01 | manufacturer/tradename Hamilton 1162-01 | manufacturer/tradename Hamilton 1162-03 |
| syringe volume 100 μL | syringe volume - | syringe volume 50 μL | syringe volume - |
Still not finding the right product?
Explore all of our products under Hamilton® Kolbenbausätze mit Spitzen für die Serien 1000, 1700 und 1800
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?