Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
Empirische Formel (Hill-System):
C2H5Cl
CAS-Nummer:
Molekulargewicht:
64.51
EC Number:
200-830-5
UNSPSC Code:
12142100
PubChem Substance ID:
Beilstein/REAXYS Number:
1730751
MDL number:
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenvapor density
2.22 (vs air)
vapor pressure
19.2 psi ( 20 °C)
assay
≥98.0%
bp
12.3 °C (lit.)
mp
−139 °C (lit.)
density
0.921 g/mL at 0-4 °C (lit.)
SMILES string
CCCl
InChI
1S/C2H5Cl/c1-2-3/h2H2,1H3
InChI key
HRYZWHHZPQKTII-UHFFFAOYSA-N
Packaging
Druckdose à 250 mL mit netto ~225 g
Other Notes
Kann Verkaufsbeschränkungen unterliegen.
Still not finding the right product?
Explore all of our products under Chlorethan
signalword
Danger
Hazard Classifications
Aquatic Chronic 3 - Carc. 2 - Flam. Gas 1 - Press. Gas Liquefied gas - Repr. 1B
Lagerklasse
2A - Gases
wgk
WGK 2
flash_point_f
-58.0 °F - closed cup
flash_point_c
-50.0 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.
Suzuko Yamazaki et al.
The journal of physical chemistry. A, 114(15), 5092-5098 (2010-03-27)
The photocatalytic degradation of chlorinated ethanes was studied in a tubular photoreactor packed with TiO(2) pellets prepared by sol-gel method. The steady-state condition was not obtained, but the deterioration in the photocatalytic activity was observed during the irradiation. Effects of
Xi-Guang Wei et al.
The Journal of organic chemistry, 75(12), 4212-4217 (2010-05-21)
The enhanced reactivity exhibited by six pseudo-alpha-bases, RC[triple bond]CZ(-) (R = H and Cl; Z = O, S, and Se) in gas-phase E2 reactions with ethyl chloride was examined at the G2(+) level. It is found that anomalous reactivity is
Peter Barnett
Pediatric emergency care, 25(6), 415-419 (2009-06-17)
Conscious sedation is used frequently to perform procedures that may be undertaken with or without minimal sedation. Fracture manipulation can be performed with minimal discomfort in the awake patient using various techniques-for example, intravenous regional anesthesia, nerve blocks, and hematoma
Global Trade Item Number
| SKU | GTIN |
|---|---|
| M6808-10G | 04061834061328 |
| M6808-50G | 04061834061335 |


