reg. compliance
EU GMP Annex 11, FDA CFR Part 11
availability
available only in Italy, Spain, Switzerland, Canada, United Kingdom, Germany, India, (Austria, Belgium, Denmak, Finland, Ireland, Netherlands, Norway, Portugal, Sweden.), North America, France, Switzerland
application(s)
industrial qc
pharmaceutical
General description
The solution includes everything you need for instant access to detailed info on every consumable in your lab.
The M Package is intended for around 20 users and includes the following:
- Identifier boxes (200 Labels each): 15
- Smart Seal Boxes (250 Labels each): 18
- Position Boxes (50 Labels each): 3
- Approximately 6 hours of user training, depending on need.
- Weekly health checks on support status and product quality.
- Quarterly and annual business review meetings with all relevant stakeholders.
Application
The iOS and Android app lets you register consumables simply by scanning them with a smartphone, and gives you access to all the inventory manager’s capabilities.(We don’t sell Android or iOS phones but can recommend a tested model.)
2. RFID Labels:
Our full range of radiofrequency identification (RFID) labels can be applied to consumables from any vendor, as well as to in-house preparations:
- Identifier labels to manage the reagents, buffers, samples and in-house prepared solutions
- Smart Seal Labels with the opening detection calculates their expiry date based on the life span
- Position labels for tracking storage locations, such as cabinets, shelves, refrigerators, freezers etc.
Web app lets you take care of your application settings and search/export functions such as restocking rules, user permissions, safety rules, audit trails export, etc.
Features and Benefits
- Vendor neutrality: You can apply the RFID labels to every consumable in your inventory, whether it was sold by Merck/Millipore Sigma or by another vendor.
- Lab personnel guidance, support, and alerts for general operational tasks and experimental workflows.
- Real-time storage compliance monitoring that alerts lab personnel to storage incompatibilities.
- Linking each consumable directly to its safety data sheet, giving users rapid electronic access to critical safety information such as safe-handling and disposal instructions.
- Greater productivity and efficiency.
- Fewer errors.
- Less waste.
- A safer workplace and environment.
- Greater compliance.
- Lower costs.