material
316L stainless steel plate, EPDM rubber O-ring, ceramic bearing (Male Bearing UNCD coating)
parameter
5-135 °C temp. range (41-275 °C)
roughness average (RA)
≤0.38 μm (Surface Treatment: Electropolished and UNCD coated)
pH range
1-14