Merck
Z671959
KNF LIQUIPORT chemically-resistant diaphragm liquid pumps
AC/DC input 115 V AC, PTFE (head), flow rate 0.5-3.0 L/min
About This Item
UNSPSC-Code:
41105101
NACRES:
NB.79
Materialien
PTFE (head)
Leistungsmerkmale
CSA approved
Hersteller/Markenname
KNF NF 300 FT.18 S
Verfügbarkeit
not available in EU
Parameter
0.5-3.0 L/min flow rate
Röhrchen ID
12 mm
Gewicht
1.5 kg
Ausgabe
24 watts (motor)
Allgemeine Beschreibung
KNF′s new NF liquid pumps are the convenient way to transfer liquids in the laboratory without the mess of peristaltic tubing. They are stylish and compact, and PTRFE models are highly resistant to chemical attack, prevent contamination f your pumped medium.
The NF100 and NF300 pumps are available in either polypropylene or PTFE head materials, with FFPM valve. You should always check corrosion compatibility charts with the medium you intend to pump.
  • 10 mWg pressure head
  • 3 mWg suction head
  • IP65 protection class
  • Maintenance-free
  • Self priming
  • Dry running
  • Adjustable flow rate
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)