Merck
Z651737
Quickfit® culture vessels
capacity 10,000 mL
Synonym(e):
FV10L
About This Item
UNSPSC-Code:
41122104
NACRES:
NB.22
Materialien
borosilicate glass
flat neck
Sterilität
non-sterile
Leistungsmerkmale
autoclavable
without cap
Verpackung
pack of 5 ea
Hersteller/Markenname
Quickfit FV10L
Verfügbarkeit
available only in EU
not available in ROW, USA and Canada
H × Durchm.
375 mm × 227 mm
Bohrungsgröße
100 mm
Kapazität
10,000 mL
Arbeitsvolumen
10000 mL
Bindungsart
non-treated surface
Allgemeine Beschreibung
  • Ideal for fermentation and culture work
  • for use with Quickfit flat-flange lids and retaining clips
Rechtliche Hinweise
Quickfit is a registered trademark of SciLabware Limited