Z532282
Aldrich® pear-shaped flask
capacity 25 mL, joint: ST/NS 14/20
Synonym(e):
pear flask, pear shape flask, round flasks
About This Item
Empfohlene Produkte
Materialien
borosilicate glass
pear-shaped flask
short
single-neck
Verbindung
joint: ST/NS14/20
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
pack of 1
Hersteller/Markenname
Sigma-Aldrich
Kapazität
25 mL