Merck
Z272965
FT-IR Spektral-Bibliothek
Nicolet®/Aldrich Vapor Phase Library
About This Item
UNSPSC-Code:
41100000
Allgemeine Beschreibung
Contains 5,010 gas phase spectra collected by Aldrich using a GC interface to ensure chromatographically pure samples. The GC interface reduces the sample decomposition common when vapor phase spectra are obtained in a conventional gas cell.
Libraries may be used with OMNIC®, GRAMS®/32, or Spectral ID® (Z407488) and are supplied on CD-ROM. Libraries are in deresolved format and contain spectra at 16 cm-1 resolution by 8-bit ordinate precision.
• Contain spectral data from 4,000 to 450 cm-1
• Best used when searching compounds with significant spectral differences
• Generally not suitable when using spectral subtraction or other precision spectral identification techniques
Sonstige Hinweise
Other libraries are available –please inquire.
• Alcohols and phenols
• Aldehydes and ketones
• Carboxylic acids, esters, lactones, and anhydrides
• Polymers
• Solvents
• More
Rechtliche Hinweise
GRAMS is a registered trademark of Thermo Electron Corp.
Nicolet is a registered trademark of Thermo Electron Corp.
OMNIC is a registered trademark of Thermo Electron Corp.
Spectral ID is a registered trademark of Galactic Industries Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)