Merck
Z147605
Sigma-Aldrich
Aldrich® bushing-type reducing adapter
female joint: ST/NS 24/40, male joint: SJ 65/40
Synonym(e):
glass adapter, glass adapters
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Verbindung
female joint: ST/NS24/40
male joint: SJ65/40
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC