Merck
WHA69702504
Whatman® GD/XP syringe filters
Nylon membrane
Synonym(e):
Whatman filter, Z338389, syringe filter, syringe filters
About This Item
UNSPSC-Code:
41104922
NACRES:
NB.24
Materialien
Nylon membrane
polypropylene housing
Sterilität
non-sterile
Leistungsmerkmale
Polypropylene prefilter
Verpackung
pack of 150 ea
Hersteller/Markenname
Whatman 6970-2504
Whatman Article No. 28420561 (US reference)
Durchmesser
25 mm
Volumen
≤25 mL
Porengröße
0.45 μm
Fitting
female Luer inlet (lock)
male Luer outlet
Leistungsmerkmale und Vorteile
  • Incorporates a two-layer polypropylene prefilter stack rather than a multi-layer glass fiber stack to minimize levels of ion extractables
  • Extremely low protein binding syringe filter
  • High loading volume for filtration of challenging samples
Eignung
Suitable for filtering trace metal analysis samples and for particulate laden sample preparation.
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva