Merck
TEM-LC300-AU-150
Sigma-Aldrich
Lacey Carbon Supported Gold Grid
size 300 mesh, hole size 150 μm , Average, box of 25
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Verpackung
box of 25
Lochgröße
150 μm , Average
Größe
300 mesh
Allgemeine Beschreibung
Lacey Carbon Supported Grid: Gold Grid, 300 mesh. Average hole size is 150 micron. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)