Merck
TEM-FF400CUUL
Sigma-Aldrich
Ultrathin Formvar Supported Copper Square Mesh
size 400 mesh, box of 50
Synonym(e):
FORMVAR SQUARE MESH - CU 400 MESH UL 50/BX
About This Item
UNSPSC-Code:
41111761
Verpackung
box of 50
Größe
400 mesh
Allgemeine Beschreibung
A 5-6 nm ultrathin formvar film supported standard copper square mesh grids. Copper grid, 400 mesh. All of our film is laid on the shiny side of the grid.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)