Merck
SYNF461100
Synthware storage flask with standard taper side arm
capacity 100 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonym(e):
Synthware F461100, round flask with side arm
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
round bottom
single-neck
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F461100
Kapazität
100 mL
Ventilgröße
0-4 mm
Allgemeine Beschreibung
Supplied with high vacuum valve with PTFE O-ring at tip.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.