SYND151819
Synthware™ short path distillation head
joint: ST/NS 19/22
Synonym(e):
Synthware D151819, Short path head
Empfohlene Produkte
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware D151819
Verwandte Kategorien
Allgemeine Beschreibung
Short-path design for microscale application, used with 25 mm immersion thermometer, and D27 distillation receiver.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.