Merck
GF47909953
Aluminiumoxid
Bolt, size m6 x 40 mm, length 25 pcs, head style hexagonal
Synonym(e):
AL60BO67
About This Item
UNSPSC-Code:
31161620
NACRES:
NA.23
Verpackung
pkg of 25
Hersteller/Markenname
Goodfellow 479-099-53