Merck
CLS6980G8
Pyrex® NMR tubes 5 mm diameter
60 MHz, L 8 in.
Synonym(e):
Corning® 6980
About This Item
UNSPSC-Code:
41121705
NACRES:
NB.43
Länge
8 in.
Allgemeine Beschreibung
These precision 5mm diameter, 8 inch long PYREX™ NMR tubes are made from quality borosilicate glass for compatibility with ground joints, valves, and vacuum racks. They are designed for use on NMR spectrometers running from 60MHz. They have an O.D. of 4.97 + 0.000-0.013mm. For replacement caps, see CLS69895.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)