Merck
CLS49956L
Pyrex® wide-mouth graduated Erlenmeyer flask with screw-cap
capacity 6,000 mL
Synonym(e):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 4995
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
polypropylene cap
screw top cap
wide-mouth
Sterilität
non-sterile
Leistungsmerkmale
cap
closure type Screw cap
Verpackung
case of 1
Hersteller/Markenname
Corning 4995-6L
Kapazität
6,000 mL
Allgemeine Beschreibung
Features a GL 45 size, PP, plug-seal screw-cap on a wide, 29 mm neck opening that accommodates larger volume serological pipettes. Also good for media prep and storage, or as a culture flask when optional membrane cap is used.
Sonstige Hinweise
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)