Merck
BR708016
Discharge tube for Dispensette® S
Organic, 25, 50 and 100 mL,
fine tip
, L 135 mm
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Materialien
polypropylene cap
Hersteller/Markenname
BRAND 708016
Verpackung
1 ea of
Länge
135 mm
Kapazität
25, 50 and 100 mL
Allgemeine Beschreibung
Without recirculation valve
Rechtliche Hinweise
Dispensette is a registered trademark of BRAND GMBH + CO KG