Merck
BR4630150
BRAND® Dispensette® S Organic, Analog-adjustable bottle-top dispenser
volume range 2.5-25 mL, without recirculation valve
Synonym(e):
bottle top dispensers
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Leistungsmerkmale
incremental 0.5 mL
DE-M marking
without recirculation valve
Hersteller/Markenname
BRAND 4630150
Volumenbereich
2.5-25 mL
Fitting
(GL 32-33)
GL 38
S 40
Allgemeine Beschreibung
Dispensette S Organic is ideal for dispensing of organic solvents and acids
Leistungsmerkmale und Vorteile
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Sonstige Hinweise
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Die Dokumentenbibliothek aufrufen
Protokolle
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Verwandter Inhalt
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.