Merck
BR29210
BRAND® soaking jar
for pipette length 460 mm
Synonym(e):
Pipette Rinsing Systems, Pipettes, Soaking jar
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Materialien
high-density polyethylene
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 29210
ID × H
150 mm × 510 mm
Kapazität
10 L
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG