Merck
BAF371110158
Spinbar® Magnetrührstäbchen
cylindrical white PTFE-coated, L × diam. 1.625 in. (41.3 mm) × 0.375 in. (9.5 mm)
Synonym(e):
Spinbar® magnetic stir bar
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.51
Materialien
cylindrical white PTFE-coated
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F37111-0158
L × Durchm.
1.625 in. (41.3 mm) × 0.375 in. (9.5 mm)
Allgemeine Beschreibung
  • Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
  • Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
  • Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
  • Autoclave safe.
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics.
Leistungsmerkmale und Vorteile
  • Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics.
  • Small removable pivot ring in the center adds to their versatility
  • Pivot ring minimizes the contact area of the bar to the vessel, reduces friction and lessens marring of plastic containers
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE® brand PTFE Coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company