Skip to Content
MilliporeSigma

T2252

apo-Transferrin human

≥95% protein basis (biuret)

Synonym(s):

Human transferrin, Siderophilin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
234-318-8
NACRES:
NA.61
MDL number:
eCl@ss:
34058011
Form:
powder
Assay:
≥95% protein basis (biuret), ≥98% (agarose gel electrophoresis)
Biological source:
human
Mol wt:
76-81 kDa
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

assay

≥95% protein basis (biuret), ≥98% (agarose gel electrophoresis)

form

powder

mol wt

76-81 kDa

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 50 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TF(7018)

General description

Transferrin is a bilobed glycoprotein containing an N-terminal lobe and a C-terminal lobe. Each of these domains is divided into two sub-domains and within the inter subdomain cleft is the metal-binding site. A single transferrin molecule can bind to two ferric ions. Apo-transferrin is an iron-free form.

Application

Apo-transferrin has been shown to strongly inhibit bacterial adhesion to biomaterials including polystyrene, polymethylmethacrylate, and silicone. Inhibition was seen at twenty micrograms/ml in culture medium.
apo-Transferrin human has been used in Dulbecco′s Modified Eagle Medium/Nutrient Mixture F-12 to culture:
  • pancreatic ductal adenocarcinoma (PDAC) cell lines
  • oligodendrocytes
  • renal proximal tubule epithelial cells (RPTEC/TERT1) cells

Biochem/physiol Actions

Transferrin transports iron through receptor-mediated endocytosis. The iron transport mechanism occurs in the endothelial cells of the blood-brain barrier. Transferrin maintains iron homeostasis and is an iron-associated marker. Reduced levels of transferrin are observed in severe COVID-19 conditions. It may bind to other molecules apart from iron, hence it can be used in targeted drug delivery.

Other Notes

Non-heme iron-transport protein.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Still not finding the right product?

Explore all of our products under apo-Transferrin human


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library