MilliporeSigma
  • Polythiol-containing, recombinant mannosylated-albumin is a superior CD68+/CD206+ Kupffer cell-targeted nanoantioxidant for treatment of two acute hepatitis models.
Polythiol-containing, recombinant mannosylated-albumin is a superior CD68+/CD206+ Kupffer cell-targeted nanoantioxidant for treatment of two acute hepatitis models.
The Journal of pharmacology and experimental therapeutics (2014-11-16)
Hitoshi Maeda, Kenshiro Hirata, Hiroshi Watanabe, Yu Ishima, Victor Tuan Giam Chuang, Kazuaki Taguchi, Akihito Inatsu, Manabu Kinoshita, Motohiko Tanaka, Yutaka Sasaki, Masaki Otagiri, Toru Maruyama
ABSTRACT
Since reactive oxygen species (ROS) derived from Kupffer cells (KC), especially CD68(+) KC, play a key role in the induction of hepatic oxidative stress and injuries, we developed a polythiolated- and mannosylated human serum albumin (SH-Man-HSA), which functions as a novel nanoantioxidant for delivering thiol to CD68(+) KC. In vitro electron paramagnetic resonance coupled with pharmacokinetics and immunohistochemical studies showed that SH-Man-HSA possessed powerful radical-scavenging activity and rapidly and selectively delivered thiols to the liver via mannose receptor (CD206) on CD68(+) cells. SH-Man-HSA significantly improved the survival rate of concanavalin-A (Con-A)-treated mice. Moreover, SH-Man-HSA exhibited excellent hepatoprotective functions, not by decreasing tumor necrosis factor or interferon-γ production that is closely associated with Con-A-induced hepatitis, but by suppressing ROS production. Interestingly, the protective effect of SH-Man-HSA was superior to N-acetyl cysteine (NAC). This could be attributed to the difference in the inhibition of hepatic oxidative stress between the two antioxidants depending on their potential for thiol delivery to the liver. Similar results were also observed for acetaminophen (APAP)-induced hepatopathy models. Flow cytometric data further confirmed that an increase in F4/80(+)/ROS(+) cells was dramatically decreased by SH-Man-HSA. The administration of SH-Man-HSA at 4 hours following a Con-A or APAP injection also exhibited a profound hepatoprotective action against these hepatitis models, whereas this was not observed for NAC. It can be concluded therefore that SH-Man-HSA has great potential for use in a rescue therapy for hepatopathy as a nanoantioxidant because of its ability to efficiently and rapidly deliver thiols to CD68(+)/CD206(+) KC.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
α-(4-Pyridyl N-oxide)-N-tert-butylnitrone, ~95%
Sigma-Aldrich
L-Glutathione oxidized disodium salt, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Alanine, European Pharmacopoeia (EP) Reference Standard
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
5,5-Dimethyl-1-pyrroline N-oxide, for ESR-spectroscopy
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
α-(4-Pyridyl N-oxide)-N-tert-butylnitrone, 99%
Sigma-Aldrich
Formaldehyde solution, 10%
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Acetaminophen, analytical standard