Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W381810

Sigma-Aldrich

DL-Alanine

≥99%, FCC, FG

Synonym(s):

(±)-2-Aminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)COOH
CAS Number:
Molecular Weight:
89.09
FEMA Number:
3818
Beilstein:
1720250
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
17.024
NACRES:
NA.21
Organoleptic:
sweet
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 172.540
FDA 21 CFR 172.54

Assay

≥99%

mp

289 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

sweet

SMILES string

CC(N)C(O)=O

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)

InChI key

QNAYBMKLOCPYGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Piezoelectric characteristics of PVA/DL-alanine polycrystals in d(33) mode.: This study explores the piezoelectric properties of polyvinyl alcohol (PVA) mixed with DL-alanine, demonstrating its potential applications in biocompatible piezoelectric devices (Jeon et al., 2023).

  • dl-Alanine Covalently Bonded Giant Arsenotungstate with Rapid Photochromic and Decent Proton Conduction Properties.: The research presents a novel DL-alanine bonded compound exhibiting significant photochromic and proton conduction capabilities, which can be leveraged in advanced materials science applications (Zheng et al., 2022).

  • Paper-based microfluidic system and chiroptical functionalized gold nano-oval for colorimetric detection of L-Tryptophan.: Although focused on L-Tryptophan, this research includes DL-alanine as a reference in developing a microfluidic system for biochemical assays (Karimian et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tatjana Braun et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(30), 9340-9345 (2015-07-01)
The prokaryotic origins of the actin cytoskeleton have been firmly established, but it has become clear that the bacterial actins form a wide variety of different filaments, different both from each other and from eukaryotic F-actin. We have used electron
Randolph M Baral et al.
Journal of feline medicine and surgery, 17(8), 653-666 (2014-10-10)
In-house analysers are commonplace in small animal practices but cannot be calibrated by the operator; therefore, any bias in the generated plasma analyte values cannot be corrected. Guidelines such as grading of renal disease and published reference intervals (RIs) in
Janine Rohrbach et al.
Antiviral therapy, 19(2), 149-159 (2013-09-17)
We investigated changes in biomarkers of liver disease in HIV-HCV-coinfected individuals during successful combination antiretroviral therapy (cART) compared to changes in biomarker levels during untreated HIV infection and to HIV-monoinfected individuals. Non-invasive biomarkers of liver disease (hyaluronic acid [HYA], aspartate
Gad Cotter et al.
European journal of heart failure, 16(12), 1362-1371 (2014-11-06)
Worsening heart failure (WHF) in the first 7 days after an admission for acute HF (AHF) has been proposed as a therapeutic target in several recent AHF studies and was a co-primary endpoint of the VERITAS studies. Patients were randomized
Veronika Fedirko et al.
Hepatology (Baltimore, Md.), 60(4), 1222-1230 (2014-03-20)
The association between vitamin D status and hepatocellular carcinoma (HCC) has not been well investigated, despite experimental evidence supporting an important role of vitamin D in liver pathophysiology. Our objective was to investigate the association between prediagnostic circulating 25-hydroxyvitamin D

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service