Skip to Content
MilliporeSigma

11-0730

Formaldehyde solution

SAJ first grade, ≥35.0%, contains methanol as stabilizer

Synonym(s):

Formalin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
PubChem Substance ID:
UNSPSC Code:
41115710
Beilstein/REAXYS Number:
1209228
MDL number:
Grade:
SAJ first grade
Bp:
97 °C ((37% solution) (lit.))
Vapor pressure:
52 mmHg ( 37 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

SAJ first grade

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

form

liquid

autoignition temp.

572 °F

contains

methanol as stabilizer

availability

available only in Japan

concentration

≥35.0%

dilution

(for analytical testing)

bp

97 °C ((37% solution) (lit.))

mp

-15 °C (lit.)

density

1.09 g/mL at 25 °C (lit.)

application(s)

sample preparation

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Formaldehyde solution


signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

target_organs

Eyes,Central nervous system, Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

147.2 °F - closed cup

flash_point_c

64 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library