MilliporeSigma
  • Synthesis and evaluation of fluorine-substituted phenyl acetate derivatives as ultra-short recovery sedative/hypnotic agents.
Synthesis and evaluation of fluorine-substituted phenyl acetate derivatives as ultra-short recovery sedative/hypnotic agents.
PloS one (2014-05-07)
Heng Zhang, Xiangqing Xu, Yin Chen, Yinli Qiu, Xin Liu, Bi-Feng Liu, Guisen Zhang
ABSTRACT
Soft drugs are molecules that are purposefully designed to be rapidly metabolized (metabolically labile). In anesthesia, the soft drug is useful because it enables precise titration to effect and rapid recovery, which might allow swift and clear-headed recovery of consciousness and early home readiness. Propofol may cause delayed awakening after prolonged infusion. Propanidid and AZD3043 have a different metabolic pathway compared to propofol, resulting in a short-acting clinical profile. Fluorine imparts a variety of properties to certain medicines, including an enhanced absorption rate and improved drug transport across the blood-brain barrier. We hypothesized that the introduction of fluorine to the frame structure of propanidid and AZD3043 would further accelerate the swift and clear-headed recovery of consciousness. To test this hypothesis, we developed a series of fluorine-containing phenyl acetate derivatives. Fluorine-containing phenyl acetate derivatives were synthesized, and their hypnotic potencies and durations of LORR following bolus or infusion administration were determined in mice, rats and rabbits. The metabolic half-lives in the blood of various species were determined chromatographically. In vitro radioligand binding and γ-aminobutyric acidA (GABAA) receptor electrophysiology studies were performed. Among the 12 synthesized fluorine-containing phenyl acetate derivatives, compound 5j induced comparable duration of LORR with AZD3043, but more rapid recovery than AZD3043, propanidid and propofol. The time of compound 5j to return to walk and behavioral recovery are approximately reduced by more than 50% compared to AZD3043 in mice and rats and rabbits. The HD50 of compound 5j decreased with increasing animal size. The rapid recovery might make compound 5j suitable for precise titration and allow swift and clear-headed recovery of consciousness and early home readiness.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Propofol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
1,1,1-Trifluoro-2-propanol, 97%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%