Skip to Content
MilliporeSigma

06473

Formic acid

greener alternative

BioUltra, 1.0 M in H2O

Synonym(s):

Methanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352107
MDL number:
Beilstein/REAXYS Number:
1209246
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Formic acid solution, BioUltra, 1.0 M in H2O

product line

BioUltra

Quality Segment

form

liquid

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

1.0 M in H2O

impurities

insoluble matter, passes filter test

refractive index

n20/D 1.336

pH

1.0-2.5 (25 °C)

cation traces

Al: ≤1 mg/kg, Ba: ≤1 mg/kg, Bi: ≤1 mg/kg, Ca: ≤5 mg/kg, Cd: ≤1 mg/kg, Co: ≤1 mg/kg, Cr: ≤1 mg/kg, Cu: ≤1 mg/kg, Fe: ≤1 mg/kg, K: ≤20 mg/kg, Li: ≤1 mg/kg, Mg: ≤1 mg/kg, Mn: ≤1 mg/kg, Mo: ≤1 mg/kg, Na: ≤20 mg/kg, Ni: ≤1 mg/kg, Pb: ≤1 mg/kg, Sr: ≤1 mg/kg, Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.03, λ: 280 nm Amax: 0.01

greener alternative category

SMILES string

[H]C(O)=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalysis. Click here for more information.

Application

Formic acid solution can be used to study biological buffers and buffer solutions. Formic acid solution has been used in a study to develop a sensitive and easy analytical method for catecholamine metabolites including 4-hydroxy-3-methoxyphenylglycol sulfate (HMPG sulfate), vanillylmandelic acid (VMA) and homovanillic acid (HVA) determination. Formic acid solution has also been used in a study to develop a method for the determination of the eight quinolones of veterinary use regulated by the European Union.


Still not finding the right product?

Explore all of our products under Formic acid solution


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter

flash_point_f

>212.0 °F

flash_point_c

> 100 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library