Skip to Content
MilliporeSigma
All Photos(1)

Documents

01-0622

Sigma-Aldrich

Acetonitrile

≥99.8%, for residue analysis, JIS 1000

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

for residue analysis

Agency

JIS 1000

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

Assay

≥99.8%

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

availability

available only in Japan

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timo Glatter et al.
Molecular systems biology, 5, 237-237 (2009-01-22)
Protein complexes represent major functional units for the execution of biological processes. Systematic affinity purification coupled with mass spectrometry (AP-MS) yielded a wealth of information on the compendium of protein complexes expressed in Saccharomyces cerevisiae. However, global AP-MS analysis of
Kangmin Chon et al.
Water research, 81, 388-397 (2015-07-05)
In this study, the changes in UV absorbance at 254 nm (UVA254) and electron donating capacity (EDC) were investigated as surrogate indicators for assessing removal of micropollutants and bromate formation during ozonation of wastewater effluents. To measure the EDC, a novel
Maria de la Paz Celorio-Mancera et al.
Insect biochemistry and molecular biology, 61, 34-45 (2015-05-03)
More than half of the proteome from mandibular glands in caterpillars is represented by chemosensory proteins. Based on sequence similarity, these proteins are putative transporters of ligands to gustatory receptors in sensory organs of insects. We sought to determine whether
Divya Subramonian et al.
Journal of proteome research, 13(9), 3905-3918 (2014-07-30)
SUMOylation is an essential posttranslational modification and regulates many cellular processes. Dysregulation of SUMOylation plays a critical role in metastasis, yet how its perturbation affects this lethal process of cancer is not well understood. We found that SUMO-2/3 modification is
Changting Xiao et al.
Diabetes, 63(7), 2394-2401 (2014-03-04)
The dipeptidyl peptidase-4 inhibitor sitagliptin, an antidiabetic agent, which lowers blood glucose levels, also reduces postprandial lipid excursion after a mixed meal. The underlying mechanism of this effect, however, is not clear. This study examined the production and clearance of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service